Learn More
Electron Microscopy Sciences Van Gieson's Stain 500 ML
SDP
Catalog No. 5031807
Encompass_Preferred
Used in Wilder Modification of Bielschowsky's Method for staining of Collagen and reticulum. Reticulum Fibers (If beading is observed and/or a dark background, replace the ammonium hydroxide silver. References: Wilder, H.C., Amer. J. Clin. Path.: 11:817, 1935, Mallory, F.B.: Pathological Techniques, Phil., W.B. Saunders & Co. c. 1938, or AFIP Manual of Hitological Staining Techniques, NY, McGraw-Hill Publ., c. 1968/3rd ed, p. 92
Catalog No. 50-318-07 Supplier Electron Microscopy Sciences Supplier No. 263925A
Used in Wilder Modification of Bielschowsky's Method for staining of Collagen and reticulum. Reticulum Fibers (If beading is observed and/or a dark background, replace the ammonium hydroxide silver. References: Wilder, H.C., Amer. J. Clin. Path.: 11:817, 1935, Mallory, F.B.: Pathological Techniques, Phil., W.B. Saunders & Co. c. 1938, or AFIP Manual of Hitological Staining Techniques, NY, McGraw-Hill Publ., c. 1968/3rd ed, p. 92