Learn More
Van Gieson Solution, 250 mL, G-Biosciences
SCI_ED
Catalog No. S001109
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
S001109 250 mL
1 options
Catalog No. S001109 Supplier G-Biosciences Supplier No. 7862241
Add to Cart
Add to Cart
Van Gieson Solution is used to stain paraffin embedded tissue specimens.
  • It is used to to identify collagen, muscle, and red blood cells.
Specifications
Quantity 250 mL
Product Type Van Gieson Stain