Learn More
Electron Microscopy Sciences Van Gieson Solution 500 ML
SDP
Catalog No. 5031864
Encompass_Preferred
Used in Hall's Method for bilirubin. References: Hall, M.J., Amer. J. Clin., 34:313 –316, 1960 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, N.Y., p. 174, c. 1968
Catalog No. 50-318-64 Supplier Electron Microscopy Sciences Supplier No. 2661102
Used in Hall's Method for bilirubin. References: Hall, M.J., Amer. J. Clin., 34:313 –316, 1960 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, N.Y., p. 174, c. 1968