Learn More
Biocare Medical, LLC Van Gogh Yellow Diluent - 100 ml
SDP
Catalog No. 5082980
Encompass_Preferred
Van Gogh Yellow Diluent is specifically formulated to enhance certain types of antibodies and may increase titers up to 2 to 4 times.
Catalog No. 50-829-80 Supplier Biocare Medical, LLC Supplier No. PD902L
Van Gogh Yellow Diluent is specifically formulated to enhance certain types of antibodies and may increase titers up to 2 to 4 times.