Learn More
MSC Van Keuren Thread Measuring Wire, 32 TPI, 2 in Long
Catalog No. 5000529253
Encompass
This thread measuring wire set is designed for precise measurement of thread pitch diameter. It features a 32 threads per inch (TPI) with a 1/32 inch pitch and a 2 inch wire length, making it suitable for various industrial applications.
  • Nominal best wire diameter of 0.018 in
  • Nominal constant of 0.0271 in
  • Set includes 3 pieces
Catalog No. 50-005-29253 Supplier MSC Supplier No. 09186479
This thread measuring wire set is designed for precise measurement of thread pitch diameter. It features a 32 threads per inch (TPI) with a 1/32 inch pitch and a 2 inch wire length, making it suitable for various industrial applications.
  • Nominal best wire diameter of 0.018 in
  • Nominal constant of 0.0271 in
  • Set includes 3 pieces