Learn More
MSC Van Keuren Thread Measuring Wire Set, 40 TPI, 0.025 in Pitch, 3 Pieces
Catalog No. 5000533106
Encompass
This thread pitch diameter measuring wire set is designed for precise measurements. It features a 40 TPI (threads per inch) and a 0.025 inch pitch, with each wire being 2 inches long. The set includes three pieces, each with a nominal best wire diameter of 0.0144 inches and a nominal constant of 0.0217 inches.
  • Measures thread pitch diameter
  • 40 threads per inch
  • 0.025 inch pitch
  • 2 inch wire length
  • 0.0144 inch nominal best wire diameter
  • 0.0217 inch nominal constant
  • Comes in a set of 3 pieces
Catalog No. 50-005-33106 Supplier MSC Supplier No. 09186453
This thread pitch diameter measuring wire set is designed for precise measurements. It features a 40 TPI (threads per inch) and a 0.025 inch pitch, with each wire being 2 inches long. The set includes three pieces, each with a nominal best wire diameter of 0.0144 inches and a nominal constant of 0.0217 inches.
  • Measures thread pitch diameter
  • 40 threads per inch
  • 0.025 inch pitch
  • 2 inch wire length
  • 0.0144 inch nominal best wire diameter
  • 0.0217 inch nominal constant
  • Comes in a set of 3 pieces