Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadate-molybdate Reagent p.a., For phosphate determination, MilliporeSigma™ Supelco™

Catalog No. 111016645 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6645 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6645 Supplier MilliporeSigma Supelco Supplier No. 94685500ML
Add to Cart
Add to Cart

Vanadate-molybdate reagent can be prepared by dissolving ammonium molybdate and ammonium vanadate in distilled water along with concentrated nitric acid.

Reagent, suitable for eliminating interfering effects by phosphate and silicate in the AAS determination of Ca and Mg
TRUSTED_SUSTAINABILITY

Specifications

MDL Number MFCD00148759
Density 1.15 g/mL (at 20°C)
Synonym VM reagent
Grade Phosphate Determination
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.