Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadate-Molybdate Reagent, for Phosphorus, LabChem™
GSA_VA

Catalog No. LC266002 Shop All LabChem Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC266002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC266002 Supplier LabChem, Inc. Supplier No. LC266002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material Vanadate-Molybdate Reagent
Color Green-Yellow
Name Note For Phosphorus
CAS Max % 83.71, 13.66, 2.5, 0.13
CAS 7647-01-0, 12054-85-2, 7803-55-6
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Linear Formula (NH4)6Mo7O24·4H2O
Quantity 1 L
Sensitivity Passes Test
Physical Form Liquid
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 1235.86
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.