Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium ICP Standard, 10,000 ppm V in 5% HNO3, Ricca Chemical
SDP

Catalog No. PV10KN50 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PV10KN-50 50 mL Natural Poly Bottle
PV10KN-100 100 mL Natural LDPE Bottle
PV10KN-500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV10KN-50 Supplier Ricca Chemical Company Supplier No. PV10KN50
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-62-2 , 7697-37-2 , 7732-18-5
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm V
CAS Min % 77.0, 20.24, 0.77
CAS Max % 80.0, 21.07, 0.8
Equivalence 1 mL = 10 mg V
Physical Form Liquid
Color Yellow
pH < 2
Type Standard
Quantity 50 mL
Chemical Name or Material Vanadium ICP Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.