Learn More
Strem, An Ascensus Company CAS# 13476-99-8. 100g. Vanadium(III) acetylacetonate, 98%. MFCD00000033
SDP
Catalog No. 5090110810
Encompass
CAS# 13476-99-8. 100g. Vanadium(III) acetylacetonate, 98%. MFCD00000033. Molecular Weight: 348.27. Molecular Formula: V(CH3COCHCOCH3)3. Color/form: brown xtl. Strem# 23-2250. http://www.strem.com/catalog/v/23-2250/
Catalog No. 50-901-10810 Supplier Strem, An Ascensus Company Supplier No. 232250100G
CAS# 13476-99-8. 100g. Vanadium(III) acetylacetonate, 98%. MFCD00000033. Molecular Weight: 348.27. Molecular Formula: V(CH3COCHCOCH3)3. Color/form: brown xtl. Strem# 23-2250. http://www.strem.com/catalog/v/23-2250/