Vanadium(III) bromide, 99.5% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 8113518

Quantity 50g

 View more versions of this product

Catalog No. AA8113518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13470-26-3
    Physical Form Crystalline
    Assay Percent Range 99.5% (metals basis)
    Molecular Formula VBr3
    Formula Weight 290.67
    MDL Number MFCD00049695
    Synonym Vanadium tribromide
    Chemical Name or Material Vanadium(III) bromide

    Specifications

    Formula Weight 290.67
    Quantity 50g
    EINECS Number 236-736-6
    Physical Form Crystalline
    Density 4.00^1^8 g/mL
    Assay Percent Range 99.5% (metals basis)
    Molecular Formula VBr3
    MDL Number MFCD00049695
    CAS 13470-26-3
    Synonym Vanadium tribromide
    Chemical Name or Material Vanadium(III) bromide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit