Learn More
Strem, An Ascensus Company CAS# 3153-26-2. 50g. Vanadium(IV) bis(acetylacetonato)oxide, 98% (Vanadyl acetylacetonate). MFCD00000032
SDP
Catalog No. 5090110807
Encompass
CAS# 3153-26-2. 50g. Vanadium(IV) bis(acetylacetonato)oxide, 98% (Vanadyl acetylacetonate). MFCD00000032. Molecular Weight: 265.16. Molecular Formula: VO(C5H7O2)2. Color/form: bluish green pwdr. Strem# 23-2202. http://www.strem.com/catalog/v/23-2202/
Catalog No. 50-901-10807 Supplier Strem, An Ascensus Company Supplier No. 23220250G
CAS# 3153-26-2. 50g. Vanadium(IV) bis(acetylacetonato)oxide, 98% (Vanadyl acetylacetonate). MFCD00000032. Molecular Weight: 265.16. Molecular Formula: VO(C5H7O2)2. Color/form: bluish green pwdr. Strem# 23-2202. http://www.strem.com/catalog/v/23-2202/