Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pfaltz & Bauer VANADIUM PENTOXIDE 99.5%

Catalog No. 507490455
Encompass

Vanadium Pentoxide 99.5%; 100g; CAS: 1314-62-1

Catalog No. 50-749-0455 Supplier Pfaltz & Bauer Supplier No. V00460100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vanadium Pentoxide 99.5%; 100g; CAS: 1314-62-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.