Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium powder, -325 mesh, 99.5% (metals basis)

Catalog No. p-4408201
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1223403 1 g
AA1223409 10 g
AA1223418 50 g
AA1223430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1223403 Supplier Thermo Scientific Chemicals Supplier No. 01223403
Only null left
Add to Cart

CAS: 7440-62-2 | V | 50.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]

Specifications

Color Silver to Gray
Melting Point 1900°C
Boiling Point 3000°C
Appearance Silver Gray powder
Density 6.11
Quantity 1 g
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99.5% (Metals basis)
Chemical Name or Material Vanadium
Show More Show Less
Hazard Category H228-H315-H319-H335: Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-171-1
RTECSNumber YW1355000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.