Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium Sulfate (Vanadyl), MP Biomedicals

Catalog No. icn219714 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219714 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219714 Supplier MP Biomedicals, Inc Supplier No. 0521971480
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27774-13-6
Molecular Formula O5SV
Molecular Weight (g/mol) 162.996
InChI Key UUUGYDOQQLOJQA-UHFFFAOYSA-L
Synonym vanadic sulfate, vanadium oxide sulphate, oxovanadium 2+ sulfate, vanadium oxysulfate, oxovanadium iv sulfate, voso4, vanadin iv oxide sulfate, vanadium, oxosulfato, vanadium, oxysulfato, vanadyl sulfate vso5
PubChem CID 34007
ChEBI CHEBI:33146
IUPAC Name oxovanadium(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].O=[V+2]

Specifications

Quantity 100 g
Formula Weight 163
Chemical Name or Material Vanadium Sulfate (Vanadyl)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.