Learn More
Strem, An Ascensus Company CAS# 1314-62-1. 100g. Vanadium(V) oxide, 98%. MFCD00011457
SDP
Catalog No. 5090114512
Encompass
CAS# 1314-62-1. 100g. Vanadium(V) oxide, 98%. MFCD00011457. Molecular Weight: 181.88. Molecular Formula: V2O5. Color/form: -60 mesh orange to brown pwdr. Strem# 93-2321. http://www.strem.com/catalog/v/93-2321/
Catalog No. 50-901-14512 Supplier Strem, An Ascensus Company Supplier No. 932321100G
CAS# 1314-62-1. 100g. Vanadium(V) oxide, 98%. MFCD00011457. Molecular Weight: 181.88. Molecular Formula: V2O5. Color/form: -60 mesh orange to brown pwdr. Strem# 93-2321. http://www.strem.com/catalog/v/93-2321/