Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vanadium(V) oxytriethoxide

Catalog No. 4707755ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-077-55ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-077-55ML Supplier Sigma Aldrich Supplier No. 4707755ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands the complexes have potential biological relevance as well as uses in catalysis.

Specifications

CAS 1686-22-2
Density 1.139 g/mL (at 25°C)
Boiling Point 125°C to 128°C
Molecular Formula C6H15O4V
Linear Formula OV(OC2H5)3
Quantity 5 mL
MDL Number MFCD00799170
Synonym Ethyl orthovanadate; Oxovanadium triethoxide; Tri(ethoxy)vanadium oxide; Triethoxyvanadyl; Triethyl vanadate; Vanadium oxytriethoxide
Molecular Weight (g/mol) 202.12
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.