Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vanadium(V) oxytriisopropoxide

Catalog No. 40492610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-492-610G 10 g
40-492-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-492-610G Supplier Sigma Aldrich Supplier No. 40492610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vanadium(V) oxytriisopropoxide is a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands the complexes have potential biological relevance as well as uses in catalysis. Vanadium (V) oxytriisopropoxide (VOTP) may be used directly as a V precursor to dope TiO2 to form a high performance anode for lithium ion batteries.

Specifications

CAS 5588-84-1
Density 1.035 g/mL (at 25°C)
Boiling Point 80°C to 82°C (2 mmHg)
Molecular Formula C9H21O4V
Refractive Index n20/D 1.479 (literature)
Linear Formula OV(OCH(CH3)2)3
Quantity 10 g
MDL Number MFCD00015017
Synonym Triisopropoxyvanadium(V) oxide; VTIP; Vanadium(V) trisisopropoxide oxide
Molecular Weight (g/mol) 244.2
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.