Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vanadium(V) oxytripropoxide

Catalog No. 47078325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-078-325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-078-325ML Supplier Sigma Aldrich Supplier No. 47078325ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vanadium (V) oxytripropoxide (VOTP) is often used as a precursor for V2O5. It may be used to prepare vanadia-silica mixed electrically conductive sol-gels for electrochemical regeneration of reduced disodium salt (NADH) for biocatalysis.

Specifications

CAS 1686-23-3
Density 1.076 g/mL (at 25°C)
Boiling Point 82°C to 83°C/0.25 mmHg (literature)
Molecular Formula C9H21O4V
Linear Formula OV(OC3H7)3
Quantity 25 mL
MDL Number MFCD00015209
Synonym Vanadium(V) tripropoxide oxide
Molecular Weight (g/mol) 244.2
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.