Learn More
Strem, An Ascensus Company CAS# 5588-84-1. 100g. Vanadium (V) tri-i-propoxy oxide, 98+% VTIP. MFCD00015017
SDP
Catalog No. 5090110822
Encompass
CAS# 5588-84-1. 100g. Vanadium (V) tri-i-propoxy oxide, 98+% VTIP. MFCD00015017. Molecular Weight: 244.20. Molecular Formula: VO(OC3H7)3. Color/form: light yellow to light green liq. Strem# 23-5000. http://www.strem.com/catalog/v/23-5000/
Catalog No. 50-901-10822 Supplier Strem, An Ascensus Company Supplier No. 235000100G
CAS# 5588-84-1. 100g. Vanadium (V) tri-i-propoxy oxide, 98+% VTIP. MFCD00015017. Molecular Weight: 244.20. Molecular Formula: VO(OC3H7)3. Color/form: light yellow to light green liq. Strem# 23-5000. http://www.strem.com/catalog/v/23-5000/