Learn More
Copper(I) cyanide, 99%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 474.0°C |
| Color | Beige-White to Green |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | 98.5% min. (ex Cu) |
| Linear Formula | CuCN |
| Fieser | 05,166; 06,146; 09,124; 16,97 |
| Merck Index | 15, 2651 |
| Solubility Information | Solubility in water: practically insoluble in water. Other solubilities: soluble in acids and alkalies, practically insoluble in alcohol |
| Formula Weight | 89.56 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal in contact with skin. Fatal if inhaled. Fatal if swallowed. Very toxic to aquatic life with long lasting effects. Contact with acids liberates very toxic gas. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Rinse mouth. |
| EINECSNumber | 208-883-6 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.