Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 100mg, Vanadyl octaethylporphine, >95%, [27860-55-5], VOO534 599.7 C36H44N4OV

Catalog No. 501456210
Encompass_Preferred

Compound: Vanadyl octaethylporphine

CAS#: 27860-55-5

MDL#: MFCD00058868

Background: A synthetic vanadyl (oxovanadium) porphyrin specialty chemical used as a reference standard in porphyrin chemistry and geochemistry, where vanadyl porphyrins are studied as biomarkers found in petroleum and oil shale.

Storage: Store at room temperature, protected from light.

Catalog No. 50-145-6210 Supplier Frontier Specialty Chemicals Supplier No. VOO534100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Vanadyl octaethylporphine

CAS#: 27860-55-5

MDL#: MFCD00058868

Background: A synthetic vanadyl (oxovanadium) porphyrin specialty chemical used as a reference standard in porphyrin chemistry and geochemistry, where vanadyl porphyrins are studied as biomarkers found in petroleum and oil shale.

Storage: Store at room temperature, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.