Learn More
Sigma Aldrich Fine Chemicals Biosciences Vanadyl ribonucleoside com
Catalog No. 502901393
Encompass
Vanadyl ribonucleoside complexes SOLUTIONon is an inhibitor for ribonuclease (RNase). Use of vanadyl ribonucleoside complexes exhibits protection from degradation by RNase in tissue genomic RNA and protein samples.
Catalog No. 50-290-1393 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9474210ML
Vanadyl ribonucleoside complexes SOLUTIONon is an inhibitor for ribonuclease (RNase). Use of vanadyl ribonucleoside complexes exhibits protection from degradation by RNase in tissue genomic RNA and protein samples.