Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum Chemical Vanadyl Sulfate, Hydrate, 97%, Spectrum™ Chemical
SDP

Catalog No. 18614651 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
20 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-651 500 g Poly Bottle
18-614-649 25 g Poly Bottle
18-614-647 100 g Poly Bottle
18-614-650 5 kg Poly Bottle
18-614-648 20 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-614-651 Supplier Spectrum Chemical Supplier No. V1020500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123334-20-3

Specifications

Melting Point 105°C
Color Blue
CAS Max % 1
Physical Form Powder, Crystals and/or Chunks
Quantity 500 g
Assay Percent Range 98.10%
Molecular Formula O5SV
MDL Number MFCD00149933
UN Number UN2931
Synonym Vanadyl Sulfate Hydrate
Solubility Information Soluble in Water
IUPAC Name Oxovanadium(2+);sulfate;hydrate
PubChem CID 24854005
Formula Weight 163 g/mol
Percent Purity 97%
Grade Reagent
Manufacturing Origin USA
Packaging Poly Bottle
Appearance Crystalline Powder or Lumps
Density 2.5 g/cm³
Assay By Titration
Chemical Name or Material Vanadyl Sulfate, Hydrate
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.