Learn More
Medchemexpress LLC Vancomycin | 1404-90-6 | >99.0% | 50 MG
SDP
Catalog No. 5000407808
Encompass_Preferred
Vancomycin is an antibiotic used for treating bacterial infections. It is a large glycopeptide compound with a molecular weight of 1450 Da. It has a unique mode of action, inhibiting the second stage of cell wall synthesis in susceptible bacteria. It is effective against many Gram-positive bacteria, including *Staphylococcus aureus*, *Staph. epidermidis*, *Str. agalactiae*, *Str. bovis*, *Str. mutans*, viridans streptococci, and enterococci. It can also be used to induce renal injury models in research.
  • Antibiotic for bacterial infections
  • Inhibits cell wall synthesis
  • Alters cell membrane permeability
  • Selectively inhibits ribonucleic acid synthesis
  • Effective against many Gram-positive bacteria
  • Useful for inducing renal injury models in research
Catalog No. 50-004-07808 Supplier Medchemexpress LLC Supplier No. HYB067150MG
Vancomycin is an antibiotic used for treating bacterial infections. It is a large glycopeptide compound with a molecular weight of 1450 Da. It has a unique mode of action, inhibiting the second stage of cell wall synthesis in susceptible bacteria. It is effective against many Gram-positive bacteria, including *Staphylococcus aureus*, *Staph. epidermidis*, *Str. agalactiae*, *Str. bovis*, *Str. mutans*, viridans streptococci, and enterococci. It can also be used to induce renal injury models in research.
  • Antibiotic for bacterial infections
  • Inhibits cell wall synthesis
  • Alters cell membrane permeability
  • Selectively inhibits ribonucleic acid synthesis
  • Effective against many Gram-positive bacteria
  • Useful for inducing renal injury models in research