Learn More
Medchemexpress LLC Vancomycin | 1404-90-6 | 99.4% | 100 MG
SDP
Catalog No. 5000409308
Encompass_Preferred
Vancomycin is an antibiotic used for treating bacterial infections. It functions by inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, Vancomycin alters cell membrane permeability and selectively inhibits ribonucleic acid synthesis. It is active against numerous Gram-positive bacteria species, including *Staphylococcus aureus*, *Staph. epidermidis*, *Str. agalactiae*, *Str. bovis*, *Str. mutans*, viridans streptococci, and enterococci.
  • Inhibits bacterial cell wall synthesis.
  • Alters cell membrane permeability.
  • Selectively inhibits ribonucleic acid synthesis.
  • Active against various Gram-positive bacteria.
  • Used for treating bacterial infections.
Catalog No. 50-004-09308 Supplier Medchemexpress LLC Supplier No. HYB0671100MG
Vancomycin is an antibiotic used for treating bacterial infections. It functions by inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, Vancomycin alters cell membrane permeability and selectively inhibits ribonucleic acid synthesis. It is active against numerous Gram-positive bacteria species, including *Staphylococcus aureus*, *Staph. epidermidis*, *Str. agalactiae*, *Str. bovis*, *Str. mutans*, viridans streptococci, and enterococci.
  • Inhibits bacterial cell wall synthesis.
  • Alters cell membrane permeability.
  • Selectively inhibits ribonucleic acid synthesis.
  • Active against various Gram-positive bacteria.
  • Used for treating bacterial infections.