Learn More
Apexbio Technology LLC Vancomycin(Synonyms: Vancocin, Vancomycin hydrochloride, Lyphocin, Vancomycine), 250mg, CAS: 1404-90-6.
SDP
Catalog No. 5000061079
Encompass_Preferred
Vancomycin (CAS 1404-90-6) is a glycopeptide antibiotic originally isolated from Streptomyces orientalis It exerts antibacterial activity primarily by inhibiting bacterial cell wall synthesis through binding to D-Ala-D-Ala termini of peptidoglycan precursors thereby preventing proper polymerization and cross-linking Vancomycin serves as an important tool in biomedical research studying bacterial resistance mechanisms and is clinically employed in managing conditions such as Clostridium difficile-associated diarrhea and enterocolitis due to methicillin-resistant Staphylococcus aureus
Catalog No. 50-000-61079 Supplier Apexbio Technology LLC Supplier No. C6417250
Vancomycin (CAS 1404-90-6) is a glycopeptide antibiotic originally isolated from Streptomyces orientalis It exerts antibacterial activity primarily by inhibiting bacterial cell wall synthesis through binding to D-Ala-D-Ala termini of peptidoglycan precursors thereby preventing proper polymerization and cross-linking Vancomycin serves as an important tool in biomedical research studying bacterial resistance mechanisms and is clinically employed in managing conditions such as Clostridium difficile-associated diarrhea and enterocolitis due to methicillin-resistant Staphylococcus aureus