Learn More
Creative Diagnostics Human Vancomycin ELISA Kit, 96 wells, sample volume 50 μL, standard range 0, 0.5, 1.5, 4.5, 13.5, 40.5 ppb, sensitivity 0.59 ng/mL
Supplier:  Creative Diagnostics DEIANJ11
Encompass
Human Vancomycin ELISA Kit, 96 wells, sample volume 50 μL, standard range 0, 0.5, 1.5, 4.5, 13.5, 40.5 ppb, sensitivity 0.59 ng/mL
Catalog No. NC2132939