Learn More
Research Products International Corp Vancomycin Hydrochloride, 1 Gram
SDP
Catalog No. 50213727
Encompass_Preferred
Vancomycin Hydrochloride is a widely used antibiotic that belongs to the glycopeptide class of antibiotics.
It is primarily effective against Gram-positive bacteria, including methicillin-resistant Staphylococcus aureus (MRSA) and vancomycin-resistant enterococci (VRE).
In the field of microbiology and research, Vancomycin Hydrochloride is used as a selective agent in culture media. It is added to growth media to inhibit the growth of Gram-positive bacteria and selectively allow the growth of Gram-negative bacteria or other microorganisms. This property makes it a valuable tool in the isolation and identification of bacteria in laboratory settings.
Catalog No. 50-213-727 Supplier Research Products International Corp Supplier No. V065001.0
Vancomycin Hydrochloride is a widely used antibiotic that belongs to the glycopeptide class of antibiotics.
It is primarily effective against Gram-positive bacteria, including methicillin-resistant Staphylococcus aureus (MRSA) and vancomycin-resistant enterococci (VRE).
In the field of microbiology and research, Vancomycin Hydrochloride is used as a selective agent in culture media. It is added to growth media to inhibit the growth of Gram-positive bacteria and selectively allow the growth of Gram-negative bacteria or other microorganisms. This property makes it a valuable tool in the isolation and identification of bacteria in laboratory settings.