Learn More
Apexbio Technology LLC Vancomycin hydrochloride 1404-93-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062247
Encompass_Preferred
Vancomycin hydrochloride is a glycopeptide antibacterial agent commonly employed in microbiological and drug resistance research due to its specific inhibitory activity Its antibacterial function derives from the inhibition of bacterial cell wall synthesis through binding directly to D-alanyl-D-alanine terminal residues of peptidoglycan precursors consequently disrupting cell-wall assembly in Gram-positive bacteria Typically vancomycin hydrochloride serves as a positive control in assays evaluating antibiotic resistance profiles bacterial susceptibility tests and screening studies related to novel glycopeptide-derivatives Reported inhibitory concentrations (IC50) vary based on bacterial strains tested generally ranging from approximately 0 5 g/mL to 2 0 g/mL under standard laboratory assay conditions
Catalog No. 50-000-62247 Supplier Apexbio Technology LLC Supplier No. B12235.1
Vancomycin hydrochloride is a glycopeptide antibacterial agent commonly employed in microbiological and drug resistance research due to its specific inhibitory activity Its antibacterial function derives from the inhibition of bacterial cell wall synthesis through binding directly to D-alanyl-D-alanine terminal residues of peptidoglycan precursors consequently disrupting cell-wall assembly in Gram-positive bacteria Typically vancomycin hydrochloride serves as a positive control in assays evaluating antibiotic resistance profiles bacterial susceptibility tests and screening studies related to novel glycopeptide-derivatives Reported inhibitory concentrations (IC50) vary based on bacterial strains tested generally ranging from approximately 0 5 g/mL to 2 0 g/mL under standard laboratory assay conditions