Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vancomycin hydrochloride | 1404-90-6 | 95.9% | 1449.25 | C66H75Cl2N9O24 | 10 MG
SDP

Catalog No. 5000299036
Encompass_Preferred

Vancomycin (Standard) is an analytical reference standard of the antibiotic vancomycin, supplied for research and analytical applications and suitable for method development and assay calibration in laboratory settings.

  • Analytical reference standard for vancomycin
  • Suitable for method development and assay calibration
  • High purity appropriate for analytical workflows
  • Supplied in a 10 mg packaging for laboratory use
  • Key identifiers: CAS 1404-90-6; formula C66H75Cl2N9O24

Catalog No. 50-002-99036 Supplier Medchemexpress LLC Supplier No. HYB0671R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vancomycin (Standard) is an analytical reference standard of the antibiotic vancomycin, supplied for research and analytical applications and suitable for method development and assay calibration in laboratory settings.

  • Analytical reference standard for vancomycin
  • Suitable for method development and assay calibration
  • High purity appropriate for analytical workflows
  • Supplied in a 10 mg packaging for laboratory use
  • Key identifiers: CAS 1404-90-6; formula C66H75Cl2N9O24

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.