Learn More
Medchemexpress LLC Vancomycin hydrochloride | 1404-93-9 | 99.4% | 1485.71 | 1 ML
SDP
Catalog No. 5000384169
Encompass_Preferred
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. Its mechanism of action involves inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, vancomycin modifies the permeability of the cell membrane and selectively prevents ribonucleic acid synthesis. This product is for research use only and has not been fully validated for medical applications.
  • Inhibits bacterial cell wall synthesis
  • Modifies cell membrane permeability
  • Selectively inhibits ribonucleic acid synthesis
  • Acts as an antibiotic
  • Relevant for autophagy research
  • Applicable in infection research
  • Used in cancer research
Catalog No. 50-003-84169 Supplier Medchemexpress LLC Supplier No. HY1736210MM/1ML
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. Its mechanism of action involves inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, vancomycin modifies the permeability of the cell membrane and selectively prevents ribonucleic acid synthesis. This product is for research use only and has not been fully validated for medical applications.
  • Inhibits bacterial cell wall synthesis
  • Modifies cell membrane permeability
  • Selectively inhibits ribonucleic acid synthesis
  • Acts as an antibiotic
  • Relevant for autophagy research
  • Applicable in infection research
  • Used in cancer research