Learn More
Medchemexpress LLC Vancomycin (hydrochloride) | 1404-93-9 | 99.4% | 1485.71 | 25 MG
SDP
Catalog No. 5000383974
Encompass_Preferred
Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis.
  • Inhibits second stage of cell wall synthesis of susceptible bacteria.
  • Alters the permeability of the cell membrane.
  • Selectively inhibits ribonucleic acid synthesis.
  • Targets bacterial, autophagy, and antibiotic pathways.
  • Used in infection and cancer research.
  • Cited in over 85 publications.
Catalog No. 50-003-83974 Supplier Medchemexpress LLC Supplier No. 25MGHY17362R
Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis.
  • Inhibits second stage of cell wall synthesis of susceptible bacteria.
  • Alters the permeability of the cell membrane.
  • Selectively inhibits ribonucleic acid synthesis.
  • Targets bacterial, autophagy, and antibiotic pathways.
  • Used in infection and cancer research.
  • Cited in over 85 publications.