Learn More
Medchemexpress LLC Vancomycin (hydrochloride) | 1404-93-9 | 99.43% | 1485.71 | 250 MG
SDP
Catalog No. 5000383913
Encompass_Preferred
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. It functions by inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, it alters cell membrane permeability and selectively inhibits ribonucleic acid synthesis.
  • Used for treating bacterial infections
  • Inhibits the second stage of cell wall synthesis
  • Alters cell membrane permeability
  • Selectively inhibits ribonucleic acid synthesis
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-003-83913 Supplier Medchemexpress LLC Supplier No. 250MGHY17362R
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. It functions by inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, it alters cell membrane permeability and selectively inhibits ribonucleic acid synthesis.
  • Used for treating bacterial infections
  • Inhibits the second stage of cell wall synthesis
  • Alters cell membrane permeability
  • Selectively inhibits ribonucleic acid synthesis
  • Suitable for laboratory centrifugation and storage