Learn More
Medchemexpress LLC Vancomycin (hydrochloride) | 1404-93-9 | 99.47% | 1450 Da | 10 G
SDP
Catalog No. 5000384562
Encompass_Preferred
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. It works by inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, Vancomycin alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis. It is a large glycopeptide compound active against many Gram-positive bacteria like Staphylococcus aureus, Staph. epidermidis, Str. agalactiae, Str. bovis, Str. mutans, viridans streptococci, and enterococci.
  • Antibiotic for the treatment of bacterial infections.
  • Inhibits the second stage of cell wall synthesis of susceptible bacteria.
  • Alters cell membrane permeability and selectively inhibits ribonucleic acid synthesis.
  • Glycopeptide antibiotic.
  • Active against various Gram-positive bacteria.
Catalog No. 50-003-84562 Supplier Medchemexpress LLC Supplier No. HY1736210G
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. It works by inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, Vancomycin alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis. It is a large glycopeptide compound active against many Gram-positive bacteria like Staphylococcus aureus, Staph. epidermidis, Str. agalactiae, Str. bovis, Str. mutans, viridans streptococci, and enterococci.
  • Antibiotic for the treatment of bacterial infections.
  • Inhibits the second stage of cell wall synthesis of susceptible bacteria.
  • Alters cell membrane permeability and selectively inhibits ribonucleic acid synthesis.
  • Glycopeptide antibiotic.
  • Active against various Gram-positive bacteria.