Learn More
Medchemexpress LLC Vancomycin (hydrochloride) | 1404-93-9 | 99.47% | 1450 Da | 250 MG
SDP
Catalog No. 5000385546
Encompass_Preferred
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. It functions by inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, vancomycin modifies the cell membrane's permeability and selectively suppresses ribonucleic acid synthesis.
  • Inhibits bacterial cell wall synthesis.
  • Alters cell membrane permeability.
  • Selectively inhibits ribonucleic acid synthesis.
  • Active against many Gram-positive bacteria, including *Staphylococcus aureus* and enterococci.
  • Glycopeptide compound.
Catalog No. 50-003-85546 Supplier Medchemexpress LLC Supplier No. 250MGHY17362
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. It functions by inhibiting the second stage of cell wall synthesis in susceptible bacteria. Additionally, vancomycin modifies the cell membrane's permeability and selectively suppresses ribonucleic acid synthesis.
  • Inhibits bacterial cell wall synthesis.
  • Alters cell membrane permeability.
  • Selectively inhibits ribonucleic acid synthesis.
  • Active against many Gram-positive bacteria, including *Staphylococcus aureus* and enterococci.
  • Glycopeptide compound.