Learn More
Medchemexpress LLC Vancomycin (hydrochloride) | 1404-93-9 | 99.5% | 1485.71 | 5 G
SDP
Catalog No. 5000385681
Encompass_Preferred
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. It functions by inhibiting the second stage of cell wall synthesis in susceptible bacteria. It also modifies cell membrane permeability and selectively inhibits ribonucleic acid synthesis.
  • Antibiotic for bacterial infections.
  • Inhibits the second stage of cell wall synthesis in susceptible bacteria.
  • Alters cell membrane permeability.
  • Selectively inhibits ribonucleic acid synthesis.
  • Active against a large number of Gram-positive bacteria, including Staphylococcus aureus, Staph. epidermidis, Str. agalactiae, Str. bovis, Str. mutans, viridans streptococci, and enterococci.
  • Glycopeptide compound with a molecular weight of 1450 Da.
Catalog No. 50-003-85681 Supplier Medchemexpress LLC Supplier No. HY173625G
Vancomycin hydrochloride is an antibiotic used for treating bacterial infections. It functions by inhibiting the second stage of cell wall synthesis in susceptible bacteria. It also modifies cell membrane permeability and selectively inhibits ribonucleic acid synthesis.
  • Antibiotic for bacterial infections.
  • Inhibits the second stage of cell wall synthesis in susceptible bacteria.
  • Alters cell membrane permeability.
  • Selectively inhibits ribonucleic acid synthesis.
  • Active against a large number of Gram-positive bacteria, including Staphylococcus aureus, Staph. epidermidis, Str. agalactiae, Str. bovis, Str. mutans, viridans streptococci, and enterococci.
  • Glycopeptide compound with a molecular weight of 1450 Da.