Learn More
Medchemexpress LLC Vancomycin hydrochloride | 1404-93-9 | 99.8% | 1485.71 | 50 MG
SDP
Catalog No. 5000384564
Encompass_Preferred
Vancomycin hydrochloride (Standard) is the analytical standard of Vancomycin hydrochloride and is intended for research and analytical applications. It is an antibiotic used for treating bacterial infections by inhibiting the second stage of cell wall synthesis in susceptible bacteria. It also modifies cell membrane permeability and selectively inhibits ribonucleic acid synthesis. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Used as an antibiotic for bacterial infections.
  • Inhibits the second stage of cell wall synthesis.
  • Modifies cell membrane permeability.
  • Selectively inhibits ribonucleic acid synthesis.
  • Suitable for experiments in HPLC, GC, and MS.
Catalog No. 50-003-84564 Supplier Medchemexpress LLC Supplier No. HY17362R50MG
Vancomycin hydrochloride (Standard) is the analytical standard of Vancomycin hydrochloride and is intended for research and analytical applications. It is an antibiotic used for treating bacterial infections by inhibiting the second stage of cell wall synthesis in susceptible bacteria. It also modifies cell membrane permeability and selectively inhibits ribonucleic acid synthesis. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Used as an antibiotic for bacterial infections.
  • Inhibits the second stage of cell wall synthesis.
  • Modifies cell membrane permeability.
  • Selectively inhibits ribonucleic acid synthesis.
  • Suitable for experiments in HPLC, GC, and MS.