Learn More
Enzo Life Sciences Vancomycin . hydrochloride (1g). CAS: 1404-93-9
SDP
Catalog No. 502009010
Encompass_Preferred
Glycopeptide antibiotic. Interferes with cell wall synthesis. Effective against Gram-positive bacteria. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9010 Supplier Enzo Life Sciences Supplier No. ALX380279G001
Glycopeptide antibiotic. Interferes with cell wall synthesis. Effective against Gram-positive bacteria. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.