Learn More
Medchemexpress LLC HY-17362 1g Medchemexpress, Vancomycin (hydrochloride) CAS:1404-93-9 Purity:>98%
SDP
Catalog No. 502029231
Encompass_Preferred
Medchemexpress, HY-17362 1g Vancomycin (hydrochloride) CAS:1404-93-9 Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9231 Supplier Medchemexpress LLC Supplier No. HY173621G
Medchemexpress, HY-17362 1g Vancomycin (hydrochloride) CAS:1404-93-9 Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.