Learn More
Sigma Aldrich Fine Chemicals Biosciences Vancomycin hydrochloride from Streptomyces orientalis, 1404-93-9, MFCD03613611, 250 mg
Catalog No. NC0370404
Encompass
Empirical Formula (Hill Notation): C66H75Cl2N9O24 · HCl, Molecular Weight: 1485.71, potency: ≥900 μg per mg (as vancomycin base), storage temp: 2-8°C.
Catalog No. NC0370404 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. V2002250MG
Empirical Formula (Hill Notation): C66H75Cl2N9O24 · HCl, Molecular Weight: 1485.71, potency: ≥900 μg per mg (as vancomycin base), storage temp: 2-8°C.