Learn More
Enzo Life Sciences Vancomycin . hydrochloride (250mg). CAS: 1404-93-9
SDP
Catalog No. 502009011
Encompass_Preferred
Glycopeptide antibiotic. Interferes with cell wall synthesis. Effective against Gram-positive bacteria. Appearance: White to light brown powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9011 Supplier Enzo Life Sciences Supplier No. ALX380279M250
Glycopeptide antibiotic. Interferes with cell wall synthesis. Effective against Gram-positive bacteria. Appearance: White to light brown powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.