Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Vancomycin hydrochloride, >900 microgram/mg

Catalog No. AC296990050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296990050 5 g Glass Bottle
AC296990010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC296990050 Supplier Thermo Scientific Chemicals Supplier No. 296990050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Vancomycin hydrochloride (>900 microgram/mg), CAS # 1404-93-9, is an antibiotic from Streptomyces orientalis, that shows antibacterial activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vancomycin hydrochloride is a glycopeptide antibiotic and antiviral from Streptomyces orientalis
  • It shows antibiotic activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains
  • Vancomycin hydrochloride inhibits bacterial mucopeptide synthesis

Applications

  • Vancomycin hydrochloride demonstrates potent activity against gram-positive bacteria
  • It has been shown in studies to be highly effective against methicillin-resistant Staphylococcus aureus (MRSA) in vitro and in vivo
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1404-93-9
Molecular Formula C66H76Cl3N9O24
Molecular Weight (g/mol) 1485.72
MDL Number MFCD03613611,MFCD03613611
InChI Key LCTORFDMHNKUSG-UHFFFAOYNA-N
PubChem CID 124080918
IUPAC Name 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
SMILES Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2

Specifications

Type Vancomycin Hydrochloride
Physical Form Powder
pH 2.5 to 4.5 (5% soln.)
Infrared Spectrum Authentic
Packaging Glass Bottle
Water 5% max.
Color Tan to White
Quantity 5 g
Solubility Information Solubility in water: 50g/L. Other solubilities: moderately soluble in diluted methanol, insoluble in higher alcohols, acetone and ether
Formula Weight 1485.73
Percent Purity ≥80.0% (vancomycin B) (HPLC), 9.0% (individual impurities) max., greater than 900μg/mg (potency, on anhydrous substance)
Chemical Name or Material Vancomycin hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.