Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vancomycin Hydrochloride, G-Biosciences
SCI_ED

Catalog No. S139837
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139837 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139837 Supplier G-Biosciences Supplier No. RC873
Add to Cart
Add to Cart

Vancomycin is a glycopeptide antibiotic that exerts its action by preventing the transfer and addition of the muramylpentapeptides that comprise the peptidoglycan.

Vancomycin is often used synergistically with cefotaxime or carbenicillin against Agrobacterium.

Chemical Identifiers

CAS 1404-93-9

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 100 g
Percent Purity 95%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.