Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vancomycin Hydrochloride MP Biomedicals

Catalog No. ICN1955402 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1955402 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1955402 Supplier MP Biomedicals, Inc Supplier No. 02195540.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycopeptide antibiotic used to block bacterial cell wall biosynthesis in Gram-positive bacteria.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123409-00-7
Molecular Formula C66H257Cl2N9O24
Molecular Weight (g/mol) 1632.721
InChI Key ARNOCQCQWLXLRG-UHFFFAOYSA-N
PubChem CID 44134997
IUPAC Name azane;N-[1-[4-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-phenoxyphenyl]ethyl]acetamide;formic acid;methane;pentadecahydrate;dihydrochloride
SMILES C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.CC(C1=CC(=C(C=C1)OC2CC(C(C(O2)CO)O)O)OC3=CC=CC=C3)NC(=O)C.C(=O)O.N.N.N.N.N.N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.Cl.Cl

Specifications

Type Vancomycin Hydrochloride
Physical Form Powder
Quantity 250 mg
Grade Not Available
Chemical Name or Material Vancomycin Hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.