Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vancomycin Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013052 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 x 250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3052 4 x 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3052 Supplier MilliporeSigma Supelco Supplier No. PHR17324X250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Vancomycin hydrochloride is a glycopeptide antibiotic, effective for the treatment of endocarditis caused by Gram-positive bacteria. It is used to inhibit the synthesis of bacterial cell wall and prevents the synthesis of peptidoglycan by binding tightly to peptides containing D-alanyl-D-alanine at the free carboxyl end. It is isolated from both Streotomyces orientalis and Nocardia lurida.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1404-93-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C66H75 Cl2N9O24 · xHCl
Synonym Vancomycin hydrochloride
Type Certified Reference Material
Quantity 4 x 250 mg
Formula Weight 1449.25 (free base basis)
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.