Learn More
Abcam Vancomycin hydrochloride, Tricyclic glycopeptide antibiotic, 1G
Catalog No. 502706785
Encompass_Preferred
MW 1449.2 Da, Purity >95%. Tricyclic glycopeptide antibiotic. Inhibits cell wall synthesis in Gram-positive bacteria. Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6785 Supplier Abcam Supplier No. AB1412241G
MW 1449.2 Da, Purity >95%. Tricyclic glycopeptide antibiotic. Inhibits cell wall synthesis in Gram-positive bacteria. Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement