Learn More
Abcam Vancomycin hydrochloride, Tricyclic glycopeptide antibiotic, 250MG
Catalog No. 502706784
Encompass_Preferred
MW 1449.2 Da, Purity >95%. Tricyclic glycopeptide antibiotic. Inhibits cell wall synthesis in Gram-positive bacteria. Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6784 Supplier Abcam Supplier No. AB141224250MG
MW 1449.2 Da, Purity >95%. Tricyclic glycopeptide antibiotic. Inhibits cell wall synthesis in Gram-positive bacteria. Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement