Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vancomycin Hydrochloride USP, Medisca™
GREENER_CHOICESDP

Catalog No. 18048134 Shop All Medisca Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-134 25 g
18-048-031 1 g
18-048-047 5 g
18-048-068 100 g
18-048-088 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-048-134 Supplier Medisca Supplier No. 027404
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1404-93-9

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Free-flowing Powder
Color Brown to White
Quantity 25 g
Percent Purity ≥ 900 μg of Vancomycin/mg
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.