Learn More
TOKU-E VANCOMYCIN SOLN 5X10ML
Catalog No. NC2068802
Encompass_Preferred
Antibiotic, Vancomycin, ReadyMade™, sterile-filtered, 100 mg/ml in DMSO. Selective agent. Useful in cell culture and plant biology.
Catalog No. NC2068802 Supplier TOKU-E Supplier No. V0405X10ML
Antibiotic, Vancomycin, ReadyMade™, sterile-filtered, 100 mg/ml in DMSO. Selective agent. Useful in cell culture and plant biology.